Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Block for MultiTherm
™
shaker
Products
Manufacturer
Package Size
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
Z755834
Block for MultiTherm
™
shaker,
Holds 35 x 1.5 mL tubes
Expand
View Pricing
Z755796
Block for MultiTherm
™
shaker,
96x 0.2 mL or 1 x PCR plate
Expand
View Pricing
Z755877
Block for MultiTherm
™
shaker,
Holds 6 x 50 mL tubes
Expand
View Pricing
Z755842
Block for MultiTherm
™
shaker,
Holds 35 x 2.0 mL tubes
Expand
View Pricing
Z755826
Block for MultiTherm
™
shaker,
15 x 0.5 mL and 20 x 1.5 mL
Expand
View Pricing
Z755869
Block for MultiTherm
™
shaker,
Holds 12 x 15 mL tubes
Expand
View Pricing
Z755885
Block for MultiTherm
™
shaker,
Expand
View Pricing
Z742458
Block for MultiTherm
™
shaker,
Holds24 x 2 mL HPLC/autosampler vials
Expand
View Pricing
Z742461
Block for MultiTherm
™
shaker,
multipurpose chamber for water, liquid, or beads
Expand
View Pricing
Z755818
Block for MultiTherm
™
shaker,
Holds 54 x 0.5 mL tubes
Expand
View Pricing
Z742456
Block for MultiTherm
™
shaker,
Holds 1 x deep well mircoplate, max. height 54 mm
Expand
View Pricing
Z755850
Block for MultiTherm
™
shaker,
Holds 24 x 12 mm tubes
Expand
View Pricing
Z742455
Block for MultiTherm
™
shaker,
Holds for 12 x 5 x tubes
Expand
View Pricing