Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Biphenyl-4,4′-dicarboxylic acid
Linear Formula:
HO
2
CC
6
H
4
C
6
H
4
CO
2
H
CAS Number:
787-70-2
Molecular Weight:
242.23
EC Number:
212-328-3
Products
Product Category
carboxylic acid monomers (1)
monomers (1)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
225266
Biphenyl-4,4′-dicarboxylic acid,
97%
Expand
View Pricing