Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BIOshell lgG 1000 Å C4, 2.7 μm HPLC Column
Products
Brand
Supelco (2)
pH Value
Agency
USP Suitability
ID (mm)
Length (cm)
Quality Segment
Pore Size (Å)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
581384-U
BIOshell lgG 1000 Å C4, 2.7 μm HPLC Column,
column L × I.D. 15 cm × 1.5 mm
Expand
View Pricing
581385-U
BIOshell lgG 1000 Å C4, 2.7 μm HPLC Column,
column L × I.D. 5 cm × 1.5 mm
Expand
View Pricing