Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
β-Estradiol
Synonym(s):
1,3,5-Estratriene-3,17β-diol, 17β-Estradiol, 3,17β-Dihydroxy-1,3,5(10)-estratriene, Dihydrofolliculin
Empirical Formula (Hill Notation):
C
18
H
24
O
2
CAS Number:
50-28-2
Molecular Weight:
272.38
EC Number:
200-023-8
Products
Product Category
hormones (3)
bioactive small molecules (1)
Brand
Sigma-Aldrich (3)
Biological Source
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Research Area
Quality Segment
Application
Package Size
Sterilization Method
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
E2758
β-Estradiol,
BioReagent, powder, suitable for cell culture
Expand
View Pricing
E2257
β-Estradiol,
powder, γ-irradiated, suitable for cell culture
Expand
View Pricing
E1132
β-Estradiol,
analytical standard
Expand
View Pricing