Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Benzyl butyl phthalate
Linear Formula:
2-[CH
3
(CH
2
)
3
O
2
C]C
6
H
4
CO
2
CH
2
C
6
H
5
CAS Number:
85-68-7
Molecular Weight:
312.36
EC Number:
201-622-7
Products
Product Category
plasticizer (2)
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
analytical standard (1)
certified reference material (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
308501
Benzyl butyl phthalate,
98%
Expand
View Pricing
36927
Benzyl butyl phthalate,
analytical standard
Expand
View Pricing
42438
Benzyl butyl phthalate,
certified reference material,
Trace
CERT
®
, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing