Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Benchmark Sprint 6H Plus Clinical Centrifuge with swing out rotor
Synonym(s):
microcentrifuge, clinical centrifuge, benchtop centrifuge
Products
Product Category
centrifuges (6)
Manufacturer
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
BMSC33036HP
Benchmark Sprint 6H Plus Clinical Centrifuge with swing out rotor,
to hold, 6 x 15 mL, AC/DC input 115 V
Expand
View Pricing
BMSC50006H
Benchmark Sprint 6H Plus Clinical Centrifuge with swing out rotor,
to hold, 6 x 10 mL, AC/DC input 115 V
Expand
View Pricing
BMSC33036HPE
Benchmark Sprint 6H Plus Clinical Centrifuge with swing out rotor,
to hold, 6 x 15 mL, AC/DC input 230 V, EuroPlug
Expand
View Pricing
BMSC33036HPEUK
Benchmark Sprint 6H Plus Clinical Centrifuge with swing out rotor,
to hold, 6 x 15 mL, AC/DC input 230 V, UK plug
Expand
View Pricing
BMSC50006HE
Benchmark Sprint 6H Plus Clinical Centrifuge with swing out rotor,
to hold, 6 x 10 mL, AC/DC input 230 V, EuroPlug
Expand
View Pricing
BMSC50006HEUK
Benchmark Sprint 6H Plus Clinical Centrifuge with swing out rotor,
to hold, 6 x 10 mL, AC/DC input 230 V, UK plug
Expand
View Pricing