Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Benchmark Centrifuge Tube
Products
Product Category
tubes (6)
Color
Feature
Manufacturer
Capacity (mL)
Diameter (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
BMSC1005T5ST
Benchmark Centrifuge Tube,
capacity 5 mL, sterile
Expand
View Pricing
BMSC1005TB5
Benchmark Centrifuge Tube,
capacity 5 mL, non-sterile, blue
Expand
View Pricing
BMSC1005TBK5
Benchmark Centrifuge Tube,
capacity 5 mL, non-sterile, opaque black
Expand
View Pricing
BMSC1005TG5
Benchmark Centrifuge Tube,
capacity 5 mL, non-sterile, green
Expand
View Pricing
BMSC1005TR5
Benchmark Centrifuge Tube,
capacity 5 mL, non-sterile, red
Expand
View Pricing
BMSC1005TY5
Benchmark Centrifuge Tube,
capacity 5 mL, non-sterile, yellow
Expand
View Pricing