Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ASTM
®
D2887 Reference Gas Oil
Synonym(s):
RGO
EC Number:
289-220-8
Products
Brand
Supelco (2)
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
analytical standard (2)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
48873
ASTM
®
D2887 Reference Gas Oil,
pkg of 6 × 1 mL
Expand
View Pricing
506419
ASTM
®
D2887 Reference Gas Oil,
analytical standard
Expand
View Pricing