Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ASTM Colour Reference Standard
Synonym(s):
ASTM Colour Reference Standard
Products
Brand
Supelco (4)
Manufacturer
Agency
Format
Application
Reference Material Type
Special Grade
certified reference material (4)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
ASTM10
ASTM Colour Reference Standard,
ASTM D 6045, D1500 (ASTM colour 1)
Expand
View Pricing
ASTM30
ASTM Colour Reference Standard,
ASTM D 6045, D1500 (ASTM colour 3)
Expand
View Pricing
ASTM05
ASTM Colour Reference Standard,
ASTM D 6045, D1500 (ASTM colour <0.5)
Expand
View Pricing
ASTM70
ASTM Colour Reference Standard,
ASTM D 6045, D1500 (ASTM colour 7)
Expand
View Pricing