
Aspartic acid

Synonym(s):
L-Aspartic acid, (S)-(+)-Aminosuccinic acid, (S)-Aminobutanedioic acid
Linear Formula:
HO2CCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
133.10
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing