Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ART
®
self-sealing barrier pipette tips
Products
Product Category
pipette tips (24)
Manufacturer
Volume Range (µL)
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-20 of 24
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
A3098
ART
®
self-sealing barrier pipette tips,
ART 200, volume range 1-200 μL, sterile
Expand
View Pricing
Z709328
ART
®
self-sealing barrier pipette tips,
volume range 100-1000 μL, sterile, ART 1000
Expand
View Pricing
Z717193
ART
®
self-sealing barrier pipette tips,
ART 1000 REACH, maximum volume 1000 μL, sterile
Expand
View Pricing
Z569755
ART
®
self-sealing barrier pipette tips,
ART 1000E, volume range 100-1000 μL, sterile
Expand
View Pricing
A2598
ART
®
self-sealing barrier pipette tips,
ART 10, volume range 0.1-10 μL, sterile
Expand
View Pricing
Z709344
ART
®
self-sealing barrier pipette tips,
ART 5000, volume range 1000-5000 μL, sterile
Expand
View Pricing
Z569763
ART
®
self-sealing barrier pipette tips,
ART 20P, volume range 0.5-20 μL, sterile
Expand
View Pricing
A9223
ART
®
self-sealing barrier pipette tips,
ART 10F, volume range 0.5-10 μL, sterile
Expand
View Pricing
Z717177
ART
®
self-sealing barrier pipette tips,
ART 20, volume range 0.5-20 μL, sterile
Expand
View Pricing
Z709182
ART
®
self-sealing barrier pipette tips,
ART 100, volume range 1-100 μL, sterile
Expand
View Pricing
Z709166
ART
®
self-sealing barrier pipette tips,
ART GEL, volume range 0.5-100 μL, sterile
Expand
View Pricing
A2723
ART
®
self-sealing barrier pipette tips,
ART 20E, volume range 0.5-10 μL, sterile
Expand
View Pricing
A2973
ART
®
self-sealing barrier pipette tips,
ART 100E, volume range 1-100 μL, sterile
Expand
View Pricing
Z709298
ART
®
self-sealing barrier pipette tips,
ART 500, volume range 50-500 μL, sterile
Expand
View Pricing
A9348
ART
®
self-sealing barrier pipette tips,
ART 1000G, volume range 100-1000 μL, sterile
Expand
View Pricing
Z569771
ART
®
self-sealing barrier pipette tips,
ART 10 Reach, volume range 0.1-10 μL, sterile
Expand
View Pricing
Z717207
ART
®
self-sealing barrier pipette tips,
ART 200G, maximum volume 200 μL, sterile
Expand
View Pricing
A9473
ART
®
self-sealing barrier pipette tips,
ART XLP, volume range 5-100 μL, sterile
Expand
View Pricing
Z709131
ART
®
self-sealing barrier pipette tips,
ART Gel 20P, volume range 0.5-20 μL, sterile
Expand
View Pricing
Z709247
ART
®
self-sealing barrier pipette tips,
ART 200U, volume range 1-200 μL, sterile
Expand
View Pricing
Page 1 of 2
Page 1 of 2