
Aripiprazole Related Compound G

Synonym(s):
7-{4-[4-(2,3-Dichlorophenyl)piperazin-1-yl]butoxy}quinolin-2(1H)-one, Dehydroaripiprazole
Empirical Formula (Hill Notation):
C23H25Cl2N3O2
CAS Number:
Molecular Weight:
446.37
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2