
Apigenin 7-glucuronide

Synonym(s):
4′,5,7-Trihydroxyflavone 7-glucuronide, Scutellarin
Empirical Formula (Hill Notation):
C21H18O11
CAS Number:
Molecular Weight:
446.36
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing