
Aniline-13C6

Synonym(s):
13C Labeled aniline, Aminobenzene-13C6, Benzeneamine-13C6, Phenylamine-13C6
Linear Formula:
13C6H5NH2
CAS Number:
Molecular Weight:
99.08
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing