Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium iron(II) sulfate hexahydrate
Synonym(s):
Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate, Mohr’s salt, Ammonium iron(II) sulfate hexahydrate
Linear Formula:
(NH
4
)
2
Fe(SO
4
)
2
· 6H
2
O
CAS Number:
7783-85-9
Molecular Weight:
392.14
EC Number:
233-151-8
Products
Product Category
salts (4)
Brand
Supelco (1)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Reagent Type
Core
Special Grade
ACS reagent (1)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
215406
Ammonium iron(II) sulfate hexahydrate,
ACS reagent, 99%
Expand
View Pricing
1.03792
Ammonium iron(II) sulfate hexahydrate,
for analysis EMSURE
®
ISO
Expand
View Pricing
F3754
Ammonium iron(II) sulfate hexahydrate,
ReagentPlus
®
, ≥98%
Expand
View Pricing
203505
Ammonium iron(II) sulfate hexahydrate,
99.997% trace metals basis
Expand
View Pricing
09719
Ammonium iron(II) sulfate hexahydrate,
BioUltra, ≥99.0% (RT)
Expand
View Pricing
F1543
Ammonium iron(II) sulfate hexahydrate,
BioXtra, ≥98%
Expand
View Pricing