Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium carbonate
Synonym(s):
Hartshorn salt
Linear Formula:
(NH
4
)
2
CO
3
CAS Number:
506-87-6
Molecular Weight:
96.09
EC Number:
208-058-0
Products
Product Category
inorganic bases (2)
bases (1)
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
ACS reagent (1)
Puriss (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
207861
Ammonium carbonate,
ACS reagent, ≥30.0% NH
3
basis
Expand
View Pricing
379999
Ammonium carbonate,
99.999% trace metals basis
Expand
View Pricing
11204
Ammonium carbonate,
puriss., meets analytical specification of NF, Ph. Franc., FCC
Expand
View Pricing