Skip to Content
Merck
Amiloride Hydrochloride Pharmaceutical Secondary Standard; Certified Reference Material

Amiloride hydrochloride

Amiloride Hydrochloride Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Amiloride hydrochloride dihydrate, N-Amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride dihydrate
Empirical Formula (Hill Notation):
C6H8ClN7O · HCl · 2H2O
CAS Number:
Molecular Weight:
302.12

Products

Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade