Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum-silicon alloy, Al99Si1
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 37
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF45010527
Aluminum-silicon alloy, Al99Si1,
foil, 50 x 50mm, 6.0mm thickness, as rolled, 99.998%
Expand
View Pricing
GF48332505
Aluminum-silicon alloy, Al99Si1,
25m wire, 0.125mm diameter, hard
Expand
View Pricing
GF45947133
Aluminum-silicon alloy, Al99Si1,
500mm rod, 18mm diameter, as drawn
Expand
View Pricing
GF59267391
Aluminum-silicon alloy, Al99Si1,
1m wire, 1mm diameter, hard
Expand
View Pricing
GF70725337
Aluminum-silicon alloy, Al99Si1,
foil, 150 x 150mm, 6.0mm thickness, as rolled, 99.998%
Expand
View Pricing
GF18174393
Aluminum-silicon alloy, Al99Si1,
12mm rod, 3mm diameter, 99.998%
Expand
View Pricing
GF00233075
Aluminum-silicon alloy, Al99Si1,
foil, 100 x 100mm, 3.0mm thickness, as rolled, 99.998%
Expand
View Pricing
GF59898245
Aluminum-silicon alloy, Al99Si1,
foil, 100 x 100mm, 2.0mm thickness, as rolled, 99.998%
Expand
View Pricing
GF62074379
Aluminum-silicon alloy, Al99Si1,
20m wire, 0.25mm diameter, hard
Expand
View Pricing
GF86204189
Aluminum-silicon alloy, Al99Si1,
100m wire, 0.05mm diameter, hard
Expand
View Pricing
GF62014003
Aluminum-silicon alloy, Al99Si1,
foil, 150 x 150mm, 2.0mm thickness, as rolled, 99.998%
Expand
View Pricing
GF73293172
Aluminum-silicon alloy, Al99Si1,
5m wire, 0.5mm diameter, hard
Expand
View Pricing
GF43407503
Aluminum-silicon alloy, Al99Si1,
10m wire, 0.05mm diameter, hard
Expand
View Pricing
GF93426879
Aluminum-silicon alloy, Al99Si1,
5m wire, 0.025mm diameter, hard
Expand
View Pricing
GF96129760
Aluminum-silicon alloy, Al99Si1,
foil, 100 x 100mm, 6.0mm thickness, as rolled, 99.998%
Expand
View Pricing
GF53682521
Aluminum-silicon alloy, Al99Si1,
foil, 150 x 150mm, 3.0mm thickness, as rolled, 99.998%
Expand
View Pricing
GF20839316
Aluminum-silicon alloy, Al99Si1,
200mm rod, 18mm diameter, as drawn
Expand
View Pricing
GF22981024
Aluminum-silicon alloy, Al99Si1,
10m wire, 1mm diameter, hard
Expand
View Pricing
GF05882766
Aluminum-silicon alloy, Al99Si1,
25m wire, 0.5mm diameter, hard
Expand
View Pricing
GF26986348
Aluminum-silicon alloy, Al99Si1,
2m wire, 0.5mm diameter, hard
Expand
View Pricing
Page 1 of 2
Page 1 of 2