Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum-silicon alloy, Al88Si12
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Particle Size (µm)
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
GF54356014
Aluminum-silicon alloy, Al88Si12,
powder, 150micron max. particle size, atomized, 100g
Expand
View Pricing
GF92936919
Aluminum-silicon alloy, Al88Si12,
powder, 150micron max. particle size, atomized, 200g
Expand
View Pricing
GF45484836
Aluminum-silicon alloy, Al88Si12,
500mm rod, 15mm diameter, extruded
Expand
View Pricing
GF85108293
Aluminum-silicon alloy, Al88Si12,
200mm rod, 15mm diameter, extruded
Expand
View Pricing
GF34235467
Aluminum-silicon alloy, Al88Si12,
100mm rod, 15mm diameter, extruded
Expand
View Pricing
GF33015978
Aluminum-silicon alloy, Al88Si12,
50mm rod, 15mm diameter, extruded
Expand
View Pricing
GF59147196
Aluminum-silicon alloy, Al88Si12,
foil, 100 x 100mm, 2.0mm thickness
Expand
View Pricing
GF93519973
Aluminum-silicon alloy, Al88Si12,
foil, 50 x 50mm, 1.0mm thickness
Expand
View Pricing
GF73530558
Aluminum-silicon alloy, Al88Si12,
foil, 50 x 50mm, 2.0mm thickness
Expand
View Pricing
GF28602503
Aluminum-silicon alloy, Al88Si12,
foil, 150 x 150mm, 1.0mm thickness
Expand
View Pricing
GF83448838
Aluminum-silicon alloy, Al88Si12,
foil, 100 x 100mm, 1.0mm thickness
Expand
View Pricing
GF07791879
Aluminum-silicon alloy, Al88Si12,
foil, 150 x 150mm, 2.0mm thickness
Expand
View Pricing