Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum nitride
Products
Manufacturer
Physical Form
Particle Size (µm)
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
GF59964395
Aluminum nitride,
sheet, thickness 2.0 mm, diameter 9.6 mm, condition translucent
Expand
View Pricing
GF43729803
Aluminum nitride,
sheet, thickness 0.64 mm, size 25 × 25 mm, condition translucent
Expand
View Pricing
GF66225090
Aluminum nitride,
sheet, thickness 0.64 mm, size 50 × 50 mm, condition translucent
Expand
View Pricing
GF14244285
Aluminum nitride,
sheet, thickness 0.125 mm, size 25 × 25 mm, condition translucent
Expand
View Pricing
GF67057320
Aluminum nitride,
powder, mean particle size (fsss) 1.4 micron, weight 20 g, purity 99.5+%
Expand
View Pricing
GF88053734
Aluminum nitride,
sheet, thickness 0.075 mm, condition translucent, size 20 × 20 mm
Expand
View Pricing
GF70740356
Aluminum nitride,
powder, mean particle size (fsss) 1.4 micron, weight 100 g, purity 99.5+%
Expand
View Pricing
GF20203546
Aluminum nitride,
powder, max. particle size 50 micron, weight 50 g, purity 99%
Expand
View Pricing
GF82165923
Aluminum nitride,
powder, mean particle size (fsss) 1.4 micron, weight 50 g, purity 99.5+%
Expand
View Pricing
GF25442535
Aluminum nitride,
powder, max. particle size 50 micron, weight 100 g, purity 99%
Expand
View Pricing
GF42544694
Aluminum nitride,
powder, mean particle size (fsss) 1.4 micron, weight 200 g, purity 99.5+%
Expand
View Pricing