Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum-nickel alloy, Al98.5Ni1.5
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
GF33850111
Aluminum-nickel alloy, Al98.5Ni1.5,
100mm rod, 14mm diameter
Expand
View Pricing
GF31264680
Aluminum-nickel alloy, Al98.5Ni1.5,
200mm rod, 14mm diameter
Expand
View Pricing
GF04864990
Aluminum-nickel alloy, Al98.5Ni1.5,
200mm rod, 12.7mm diameter
Expand
View Pricing
GF27240511
Aluminum-nickel alloy, Al98.5Ni1.5,
200mm rod, 8mm diameter
Expand
View Pricing
GF08085359
Aluminum-nickel alloy, Al98.5Ni1.5,
500mm rod, 14mm diameter
Expand
View Pricing
GF24038917
Aluminum-nickel alloy, Al98.5Ni1.5,
100mm rod, 12.7mm diameter
Expand
View Pricing
GF05063936
Aluminum-nickel alloy, Al98.5Ni1.5,
500mm rod, 8mm diameter
Expand
View Pricing
GF10669642
Aluminum-nickel alloy, Al98.5Ni1.5,
1000mm rod, 14mm diameter
Expand
View Pricing
GF32992319
Aluminum-nickel alloy, Al98.5Ni1.5,
1000mm rod, 8mm diameter
Expand
View Pricing