Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 25
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF27304163
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
50m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF46598619
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
20m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF94177749
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
5m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF14793385
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
20m wire, 0.125mm diameter, as drawn
Expand
View Pricing
GF72470662
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
100m wire, 0.05mm diameter, as drawn
Expand
View Pricing
GF32825123
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
20m wire, 0.025mm diameter, as drawn
Expand
View Pricing
GF20145787
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
20m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF24827452
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
10m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF59525185
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
5m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF33379768
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
50m wire, 0.05mm diameter, as drawn
Expand
View Pricing
GF26187475
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
10m wire, 0.05mm diameter, as drawn
Expand
View Pricing
GF85062607
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
10m wire, 0.025mm diameter, as drawn
Expand
View Pricing
GF47880502
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
5m wire, 0.025mm diameter, as drawn
Expand
View Pricing
GF00529253
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
50m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF28961163
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
10m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF72908837
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
5m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF49172788
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
10m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF43110212
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
5m wire, 0.125mm diameter, as drawn
Expand
View Pricing
GF73434540
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
20m wire, 0.05mm diameter, as drawn
Expand
View Pricing
GF97039446
Aluminum-magnesium-silicon alloy, Al98Mg1Si0.6,
50m wire, 0.125mm diameter, as drawn
Expand
View Pricing
Page 1 of 2
Page 1 of 2