Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum/Magnesium/Silicon
Products
Manufacturer
Physical Form
Length (mm)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GF71799474
Aluminum/Magnesium/Silicon,
rod, 3.18 mm diameter, Al98.8%/Mg 0.7%/Si 0.5%, length 100 mm, temper soft
Expand
View Pricing
GF66755560
Aluminum/Magnesium/Silicon,
rod, Al98.8%/Mg 0.7%/Si 0.5%, 3.18 mm diameter, length 500 mm, temper soft
Expand
View Pricing
GF55640987
Aluminum/Magnesium/Silicon,
rod, Al 98.8%/Mg 0.7%/Si 0.5%, 4.76 mm diameter, length 100 mm, temper soft
Expand
View Pricing
GF55123602
Aluminum/Magnesium/Silicon,
rod, Al 98.8%/Mg 0.7%/Si 0.5%, 4.76 mm diameter, length 200 mm, temper soft
Expand
View Pricing
GF74934041
Aluminum/Magnesium/Silicon,
rod, 3.18 mm diameter, Al98.8%/Mg 0.7%/Si 0.5%, length 200 mm, temper soft
Expand
View Pricing
GF90451087
Aluminum/Magnesium/Silicon,
rod, 4.76 mm diameter, Al98.8%/Mg 0.7%/Si 0.5%, length 500 mm, temper soft
Expand
View Pricing
GF06385833
Aluminum/Magnesium/Silicon,
rod, Al 98.8%/Mg 0.7%/Si 0.5%, 4.76 mm diameter, length 1000 mm, temper soft
Expand
View Pricing
GF44445124
Aluminum/Magnesium/Silicon,
rod, Al 98.8%/Mg 0.7%/Si 0.5%, 3.18 mm diameter, length 1000 mm, temper soft
Expand
View Pricing