Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum crimp seal (without septum), silver aluminum
Products
Product Category
sealing films and foils (4)
Brand
Supelco (4)
Diameter (mm)
OD (mm)
Color
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
27016
Aluminum crimp seal (without septum), silver aluminum,
tear-away, diam. 20 mm, pkg of 100 ea
Expand
View Pricing
27230-U
Aluminum crimp seal (without septum), silver aluminum,
open center, diam. × opening 20 mm × 9.5 mm, pkg of 100 ea
Expand
View Pricing
27227-U
Aluminum crimp seal (without septum), silver aluminum,
removable center
, diam. 20 mm, pkg of 100 ea
Expand
View Pricing
27200
Aluminum crimp seal (without septum), silver aluminum,
pressure release
, diam. × opening 20 mm × 8 mm, pkg of × 100 ea
Expand
View Pricing