Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum-cobalt alloy, Al99.5Co0.5
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
GF50344725
Aluminum-cobalt alloy, Al99.5Co0.5,
5m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF20541709
Aluminum-cobalt alloy, Al99.5Co0.5,
10m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF33049762
Aluminum-cobalt alloy, Al99.5Co0.5,
0.5m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF99200976
Aluminum-cobalt alloy, Al99.5Co0.5,
1m wire, 1.0mm diameter, as drawn
Expand
View Pricing