Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum alloy 5019
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
GF30882551
Aluminum alloy 5019,
wire, Al 95%/Mg 5%, 0.08 mm diameter, length 50 m, temper hard
Expand
View Pricing
GF15828813
Aluminum alloy 5019,
wire, Al 95%/Mg 5%, 0.08 mm diameter, length 100 m, temper hard
Expand
View Pricing
GF04718910
Aluminum alloy 5019,
wire, Al 95%/Mg 5%, 0.05 mm diameter, length 50 m, temper hard
Expand
View Pricing
GF01298279
Aluminum alloy 5019,
wire, Al 95%/Mg 5%, 0.05 mm diameter, length 20 m, temper hard
Expand
View Pricing
GF08647465
Aluminum alloy 5019,
wire, Al 95%/Mg 5%, 0.12 mm diameter, length 200 m, temper hard
Expand
View Pricing
GF83619063
Aluminum alloy 5019,
wire, Al95%/Mg 5%, 0.08 mm diameter, length 10 m, temper hard
Expand
View Pricing
GF74625048
Aluminum alloy 5019,
wire, 0.08 mm diameter, Al95%/Mg 5%, length 20 m, temper hard
Expand
View Pricing
GF51227827
Aluminum alloy 5019,
wire, Al 95%/Mg 5%, 0.12 mm diameter, length 500 m, temper hard
Expand
View Pricing
GF23417782
Aluminum alloy 5019,
wire, Al 95%/Mg 5%, 0.12 mm diameter, length 100 m, temper hard
Expand
View Pricing
GF90309944
Aluminum alloy 5019,
wire, Al95%/Mg 5%, length 500 m, 0.08 mm diameter, temper hard
Expand
View Pricing
GF02792684
Aluminum alloy 5019,
wire, Al 95%/Mg 5%, 0.05 mm diameter, length 200 m, temper hard
Expand
View Pricing
GF10361214
Aluminum alloy 5019,
wire, Al 95%/Mg 5%, 0.12 mm diameter, length 50 m, temper hard
Expand
View Pricing
GF60962788
Aluminum alloy 5019,
wire, Al95%/Mg 5%, 0.05 mm diameter, length 10 m, temper hard
Expand
View Pricing
GF63014712
Aluminum alloy 5019,
wire, Al95%/Mg 5%, length 200 m, 0.08 mm diameter, temper hard
Expand
View Pricing
GF82123589
Aluminum alloy 5019,
wire, Al95%/Mg 5%, 0.05 mm diameter, length 100 m, temper hard
Expand
View Pricing
GF94230446
Aluminum alloy 5019,
wire, Al95%/Mg 5%, length 10 m, 0.12 mm diameter, temper hard
Expand
View Pricing