
Aluminium chloride hexahydrate

Synonym(s):
Aluminium chloride hexahydrate
Empirical Formula (Hill Notation):
AlCl3 · 6H2O
CAS Number:
Molecular Weight:
241.43
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing