Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminium alloy 2024
Products
Manufacturer
Physical Form
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
GF11054225
Aluminium alloy 2024,
foil, Al 93.5%/Cu 4.5%/Mg 1.5%/Mn 0.5%, thickness 2.5 mm, size 150 × 150 mm, temper t3
Expand
View Pricing
GF31514420
Aluminium alloy 2024,
foil, Al 93.5%/Cu 4.5%/Mg 1.5%/Mn 0.5%, thickness 3.0 mm, size 150 × 150 mm, temper t3
Expand
View Pricing
GF05260722
Aluminium alloy 2024,
foil, Al 93.5%/Cu 4.5%/Mg 1.5%/Mn 0.5%, thickness 3.0 mm, size 300 × 300 mm, temper t3
Expand
View Pricing
GF79405454
Aluminium alloy 2024,
foil, Al93.5%/Cu 4.5%/Mg 1.5%/Mn 0.5%, thickness 2.5 mm, size 300 × 300 mm, temper t3
Expand
View Pricing