Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
vacuum trap
Products
Brand
Sigma-Aldrich (9)
OD (mm)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z549460
Aldrich
®
vacuum trap,
Inlet, 90° bend, O.D. × H 45 mm × 300 mm
Expand
View Pricing
Z225347
Aldrich
®
vacuum trap,
Inlet, straight, O.D. × H 13 mm × 75 mm
Expand
View Pricing
Z549436
Aldrich
®
vacuum trap,
Inlet, 90° bend, O.D. × H 25 mm × 150 mm
Expand
View Pricing
Z549363
Aldrich
®
vacuum trap,
Inlet, 90
°
bend, O.D. × H 13 mm × 75 mm
Expand
View Pricing
Z549401
Aldrich
®
vacuum trap,
Inlet, straight, O.D. × H 35 mm × 250 mm
Expand
View Pricing
Z549398
Aldrich
®
vacuum trap,
Inlet, straight, O.D. × H 25 mm × 200 mm
Expand
View Pricing
Z549444
Aldrich
®
vacuum trap,
Inlet, 90° bend, O.D. × H 25 mm × 200 mm
Expand
View Pricing
Z549452
Aldrich
®
vacuum trap,
Inlet, 90° bend, O.D. × H 35 mm × 250 mm
Expand
View Pricing
Z549428
Aldrich
®
vacuum trap,
Inlet, straight, O.D. × H 45 mm × 300 mm
Expand
View Pricing