Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
two-neck round-bottom flask
Products
Product Category
flasks (10)
Brand
Sigma-Aldrich (9)
Manufacturer
Capacity (mL)
Closure Type
Package Size
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z516902
Aldrich
®
two-neck round-bottom flask,
capacity 500 mL, Joints: ST/NS 24/40 (2)
Expand
View Pricing
Z516856
Aldrich
®
two-neck round-bottom flask,
capacity 100 mL, Joints: ST/NS 24/40 (2)
Expand
View Pricing
Z516821
Aldrich
®
two-neck round-bottom flask,
capacity 50 mL, Joints: ST/NS 14/20 (2)
Expand
View Pricing
Z516864
Aldrich
®
two-neck round-bottom flask,
capacity 250 mL, Joints: ST/NS 14/20 (2)
Expand
View Pricing
Z516872
Aldrich
®
two-neck round-bottom flask,
capacity 250 mL, Joints: ST/NS 24/40 (2)
Expand
View Pricing
Z516945
Aldrich
®
two-neck round-bottom flask,
capacity 1,000 mL, Joints: ST/NS 29/32 (2)
Expand
View Pricing
Z516848
Aldrich
®
two-neck round-bottom flask,
capacity 100 mL, Joints: ST/NS 14/20 (2)
Expand
View Pricing
Z516880
Aldrich
®
two-neck round-bottom flask,
capacity 250 mL, Joints: ST/NS 29/32 (2)
Expand
View Pricing
Z516910
Aldrich
®
two-neck round-bottom flask,
capacity 500 mL, Joints: ST/NS 29/32 (2)
Expand
View Pricing
Z516937
Aldrich
®
two-neck round-bottom flask,
capacity 1,000 mL, Joints: ST/NS 24/40 (2)
Expand
View Pricing