Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
sleeve stopper septa, fluoroelastomer FKM
Products
Product Category
septa (2)
Brand
Sigma-Aldrich (2)
Color
ID (mm)
OD (mm)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
Z569984
Aldrich
®
sleeve stopper septa, fluoroelastomer FKM,
bottom I.D. × O.D. 12.7 mm × 23.7 mm, black, fits 24/40 joints and 64, 80 oz and 1gal jugs
Expand
View Pricing
Z569941
Aldrich
®
sleeve stopper septa, fluoroelastomer FKM,
bottom I.D. × O.D. 4.2 mm × 10.7 mm, black, fits 10/30 joints
Expand
View Pricing