Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
Essentials volumetric pipette, class A, meets ASTM E 969
Products
Product Category
pipettes (5)
Brand
Sigma-Aldrich (5)
Color
Manufacturer
Capacity (mL)
Glassware Class
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z740920
Aldrich
®
Essentials volumetric pipette, class A, meets ASTM E 969,
capacity 1.0 mL, tol. 0.006 mL, blue
Expand
View Pricing
Z740923
Aldrich
®
Essentials volumetric pipette, class A, meets ASTM E 969,
capacity 5 mL, tol. 0.010 mL, white
Expand
View Pricing
Z740925
Aldrich
®
Essentials volumetric pipette, class A, meets ASTM E 969,
capacity 20 mL, tol. 0.030 mL, yellow
Expand
View Pricing
Z740924
Aldrich
®
Essentials volumetric pipette, class A, meets ASTM E 969,
capacity 10 mL, tol. 0.020 mL, red
Expand
View Pricing
Z740921
Aldrich
®
Essentials volumetric pipette, class A, meets ASTM E 969,
capacity 2.0 mL, tol. 0.006 mL, orange
Expand
View Pricing