Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
desiccator cabinet
Products
Product Category
desiccators (6)
Brand
Sigma-Aldrich (6)
Color
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z268135
Aldrich
®
desiccator cabinet,
H × W × D 7 in. × 7 in. × 10 in., 2 trays, clear
Expand
View Pricing
Z268151
Aldrich
®
desiccator cabinet,
H × W × D 12 in. × 12 in. × 12 in., 3 trays, clear
Expand
View Pricing
Z268143
Aldrich
®
desiccator cabinet,
H × W × D 9 in. × 9 in. × 16 in., 2 trays, clear
Expand
View Pricing
Z268186
Aldrich
®
desiccator cabinet,
H × W × D 9 in. × 9 in. × 16 in., 2 trays, bronze
Expand
View Pricing
Z268194
Aldrich
®
desiccator cabinet,
H × W × D 12 in. × 12 in. × 12 in., 3 trays, bronze
Expand
View Pricing
Z268178
Aldrich
®
desiccator cabinet,
H × W × D 7 in. × 7 in. × 10 in., 2 trays, bronze
Expand
View Pricing