Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
corrosive lecture-bottle gas regulator
Synonym(s):
Aldrich
®
corrosive lecture-bottle gas regulator
Products
Product Category
regulators (6)
Brand
Sigma-Aldrich (5)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z148512
Aldrich
®
corrosive lecture-bottle gas regulator,
delivery pressure 1-50 psig, CGA Inlet size 180
Expand
View Pricing
Z148504
Aldrich
®
corrosive lecture-bottle gas regulator,
delivery pressure 1-15 psig, CGA Inlet size 180
Expand
View Pricing
Z512966
Aldrich
®
corrosive lecture-bottle gas regulator,
delivery pressure 0-15 psig, CGA Inlet size 170
Expand
View Pricing
Z512974
Aldrich
®
corrosive lecture-bottle gas regulator,
delivery pressure 0-50 psig, CGA Inlet size 170
Expand
View Pricing
Z742164
Aldrich
®
corrosive lecture-bottle gas regulator,
DIN 1 connection
Expand
View Pricing
Z742162
Aldrich
®
corrosive lecture-bottle gas regulator,
DIN 1 connection
Expand
View Pricing