Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
ColorSpec
®
NMR tubes
Products
Product Category
tubes (19)
Brand
Sigma-Aldrich (18)
Color
Diameter (mm)
Length (in)
Color
Frequency (MHz)
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
Z569194
Aldrich
®
ColorSpec
®
NMR tubes,
tube diam. 5 mm, size 7 in., disposable, color-coded blue, frequency 200 MHz
Expand
View Pricing
Z569348
Aldrich
®
ColorSpec
®
NMR tubes,
tube diam. × L 5 mm × 7 in., precision, color-coded red, frequency 400 MHz
Expand
View Pricing
Z569208
Aldrich
®
ColorSpec
®
NMR tubes,
tube diam. 5 mm, size 8 in., disposable, color-coded blue, frequency 200 MHz
Expand
View Pricing
Z569216
Aldrich
®
ColorSpec
®
NMR tubes,
tube diam. × L 5 mm × 7 in., thrift, color-coded yellow, frequency 200 MHz
Expand
View Pricing
Z569259
Aldrich
®
ColorSpec
®
NMR tubes,
thrift, color-coded yellow, frequency 400 MHz
Expand
View Pricing
Z569291
Aldrich
®
ColorSpec
®
NMR tubes,
tube diam. × L 5 mm × 7 in., precision, color-coded red, frequency 100 MHz
Expand
View Pricing
Z569232
Aldrich
®
ColorSpec
®
NMR tubes,
thrift, color-coded yellow, frequency 300 MHz
Expand
View Pricing
Z569224
Aldrich
®
ColorSpec
®
NMR tubes,
tube diam. 5 mm, L 8 in., thrift, color-coded yellow, frequency 200 MHz
Expand
View Pricing
Z569380
Aldrich
®
ColorSpec
®
NMR tubes,
tube diam. × L 5 mm × 7 in., precision, color-coded red, frequency 600 MHz
Expand
View Pricing
Z569364
Aldrich
®
ColorSpec
®
NMR tubes,
tube diam. × L 5 mm × 7 in., precision, color-coded red, frequency 500 MHz
Expand
View Pricing
Z569275
Aldrich
®
ColorSpec
®
NMR tubes,
tube diam. × L 5 mm × 7 in., thrift, color-coded yellow, frequency 500 MHz
Expand
View Pricing
Z569356
Aldrich
®
ColorSpec
®
NMR tubes,
tube diam. × L 5 mm × 8 in., precision, color-coded red, frequency 400 MHz
Expand
View Pricing
Z569240
Aldrich
®
ColorSpec
®
NMR tubes,
thrift, color-coded yellow, frequency 300 MHz
Expand
View Pricing
Z569283
Aldrich
®
ColorSpec
®
NMR tubes,
tube diam. × L 5 mm × 8 in., thrift, color-coded yellow, frequency 500 MHz
Expand
View Pricing
Z569399
Aldrich
®
ColorSpec
®
NMR tubes,
tube diam. × L 5 mm × 8 in., precision, color-coded red, frequency 600 MHz
Expand
View Pricing
Z569372
Aldrich
®
ColorSpec
®
NMR tubes,
tube diam. × L 5 mm × 8 in., precision, color-coded red, frequency 500 MHz
Expand
View Pricing
Z569305
Aldrich
®
ColorSpec
®
NMR tubes,
tube diam. × L 5 mm × 8 in., precision, color-coded red, frequency 100 MHz
Expand
View Pricing
Z569402
Aldrich
®
ColorSpec
®
NMR tubes,
tube diam. × L 5 mm × 7 in., precision, color-coded red, frequency 800 MHz
Expand
View Pricing
Z569267
Aldrich
®
ColorSpec
®
NMR tubes,
tube diam. × L 5 mm × 8 in., thrift, color-coded yellow, frequency 400 MHz
Expand
View Pricing