Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
coiled condenser
Products
Brand
Sigma-Aldrich (13)
Length (mm)
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
Z531073
Aldrich
®
coiled condenser,
unjacketed, L 100 mm, joint: ST/NS 14/20
Expand
View Pricing
Z531189
Aldrich
®
coiled condenser,
unjacketed, L 500 mm, joint: ST/NS 24/40
Expand
View Pricing
Z531138
Aldrich
®
coiled condenser,
unjacketed, L 300 mm, joint: ST/NS 14/20
Expand
View Pricing
Z531308
Aldrich
®
coiled condenser,
jacketed, L 400 mm, joint: ST/NS 24/40
Expand
View Pricing
Z531081
Aldrich
®
coiled condenser,
unjacketed, L 200 mm, joint: ST/NS 14/20
Expand
View Pricing
Z531227
Aldrich
®
coiled condenser,
jacketed, L 100 mm, joint: ST/NS 14/20
Expand
View Pricing
Z531154
Aldrich
®
coiled condenser,
unjacketed, L 300 mm, joint: ST/NS 29/32
Expand
View Pricing
Z531294
Aldrich
®
coiled condenser,
jacketed, L 300 mm, joint: ST/NS 29/32
Expand
View Pricing
Z531235
Aldrich
®
coiled condenser,
jacketed, L 200 mm, joint: ST/NS 14/20
Expand
View Pricing
Z531286
Aldrich
®
coiled condenser,
jacketed, L 300 mm, joint: ST/NS 24/40
Expand
View Pricing
Z531103
Aldrich
®
coiled condenser,
unjacketed, L 200 mm, joint: ST/NS 24/40
Expand
View Pricing
Z531146
Aldrich
®
coiled condenser,
unjacketed, L 300 mm, joint: ST/NS 24/40
Expand
View Pricing
Z531162
Aldrich
®
coiled condenser,
unjacketed, L 400 mm, joint: ST/NS 24/40
Expand
View Pricing