Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
calibrated chromatography column with solvent reservoir and fritted disc
Products
Product Category
columns (11)
Brand
Sigma-Aldrich (10)
OD (mm)
Volume (ml)
Technique
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
Z560367
Aldrich
®
calibrated chromatography column with solvent reservoir and fritted disc,
reservoir volume 250 mL, top joint: threaded 24/40
Expand
View Pricing
Z560405
Aldrich
®
calibrated chromatography column with solvent reservoir and fritted disc,
reservoir volume 500 mL, top joint: threaded 24/40
Expand
View Pricing
Z560340
Aldrich
®
calibrated chromatography column with solvent reservoir and fritted disc,
reservoir volume 100 mL, top joint: threaded 24/40
Expand
View Pricing
Z560359
Aldrich
®
calibrated chromatography column with solvent reservoir and fritted disc,
reservoir volume 100 mL, top joint: threaded 29/32
Expand
View Pricing
Z560375
Aldrich
®
calibrated chromatography column with solvent reservoir and fritted disc,
reservoir volume 250 mL, top joint: threaded 29/32
Expand
View Pricing
Z560464
Aldrich
®
calibrated chromatography column with solvent reservoir and fritted disc,
reservoir volume 2000 mL, top joint: threaded 45/50, column volume 2000 mL
Expand
View Pricing
Z560391
Aldrich
®
calibrated chromatography column with solvent reservoir and fritted disc,
reservoir volume 500 mL, top joint: threaded 29/32
Expand
View Pricing
Z560421
Aldrich
®
calibrated chromatography column with solvent reservoir and fritted disc,
reservoir volume 1000 mL, top joint: threaded 45/40
Expand
View Pricing
Z560456
Aldrich
®
calibrated chromatography column with solvent reservoir and fritted disc,
reservoir volume 2000 mL, top joint: threaded 45/40
Expand
View Pricing
Z560413
Aldrich
®
calibrated chromatography column with solvent reservoir and fritted disc,
reservoir volume 500 mL, top joint: threaded 29/32
Expand
View Pricing
Z560448
Aldrich
®
calibrated chromatography column with solvent reservoir and fritted disc,
reservoir volume 1000 mL, top joint: threaded 45/50
Expand
View Pricing