Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Albumin from porcine serum
CAS Number:
9048-46-8
EC Number:
232-936-2
Products
Product Category
albumins (3)
Brand
Sigma-Aldrich (3)
Biological Source
Feature
Physical Form
Purity
Quality Segment
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A1830
Albumin from porcine serum,
lyophilized powder, ≥98% (agarose gel electrophoresis)
Expand
View Pricing
A4414
Albumin from porcine serum,
lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Expand
View Pricing
A2764
Albumin from porcine serum,
lyophilized powder, 96-99% (agarose gel electrophoresis)
Expand
View Pricing