Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Albumin from human serum
Synonym(s):
HSA
CAS Number:
70024-90-7
EC Number:
274-272-6
Products
Product Category
albumins (7)
proteins (7)
Brand
Sigma-Aldrich (7)
Biological Source
Gene Alias
Physical Form
Purity
Quality Segment
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
A1653
Albumin from human serum,
lyophilized powder, ≥96% (agarose gel electrophoresis)
Expand
View Pricing
A3782
Albumin from human serum,
lyophilized powder, Fatty acid free, Globulin free, ≥99% (agarose gel electrophoresis)
Expand
View Pricing
A8763
Albumin from human serum,
lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Expand
View Pricing
A9511
Albumin from human serum,
lyophilized powder, ≥97% (agarose gel electrophoresis)
Expand
View Pricing
A1887
Albumin from human serum,
lyophilized powder, essentially fatty acid free
Expand
View Pricing
A5843
Albumin from human serum,
lyophilized powder, low endotoxin
Expand
View Pricing
A4327
Albumin from human serum,
lyophilized powder, essentially protease free, ≥96% (agarose gel electrophoresis)
Expand
View Pricing