Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Albumin from chicken egg white
Synonym(s):
Albumin from hen egg white, Ovalbumin
CAS Number:
9006-59-1
EC Number:
232-692-7
Products
Product Category
albumins (6)
proteins (5)
Brand
Sigma-Aldrich (5)
Supelco (1)
Biological Source
Physical Form
Purity
Tissue
Quality Segment
Package Size
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
A5503
Albumin from chicken egg white,
lyophilized powder, ≥98% (agarose gel electrophoresis)
Expand
View Pricing
A2512
Albumin from chicken egg white,
lyophilized powder, ≥98% (agarose gel electrophoresis)
Expand
View Pricing
A5253
Albumin from chicken egg white,
powder, 62-88% (agarose gel electrophoresis)
Expand
View Pricing
A5378
Albumin from chicken egg white,
lyophilized powder, ≥90% (agarose gel electrophoresis)
Expand
View Pricing
A7641
Albumin from chicken egg white,
lyophilized powder
Expand
View Pricing
A7642
Albumin from chicken egg white,
For use as a marker in SDS-PAGE
Expand
View Pricing