Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Adenosine 5′-triphosphate disodium salt trihydrate
Empirical Formula (Hill Notation):
C
10
H
20
N
5
Na
2
O
16
P
3
CAS Number:
51963-61-2
Molecular Weight:
605.19
EC Number:
213-579-1
Products
Formula Weight
Manufacturer
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
ATPD-RO
Adenosine 5′-triphosphate disodium salt trihydrate,
Disodium salt
Expand
View Pricing
ATPDS-RO
Adenosine 5′-triphosphate disodium salt trihydrate,
=99%, pkg of 1 g (10519979001), pkg of 5 g (10519987001)
Expand
View Pricing
797189
Adenosine 5′-triphosphate disodium salt trihydrate,
Expand
View Pricing