![Adenine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/767/131/7b4057e4-5d02-4ed4-9bd1-553c61f97782/640/7b4057e4-5d02-4ed4-9bd1-553c61f97782.png)
Adenine
![Adenine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/767/131/7b4057e4-5d02-4ed4-9bd1-553c61f97782/640/7b4057e4-5d02-4ed4-9bd1-553c61f97782.png)
Synonym(s):
6-Aminopurine, Vitamin B4, Adenine
Empirical Formula (Hill Notation):
C5H5N5
CAS Number:
Molecular Weight:
135.13
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing