Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Adapter for Eppendorf
®
FA-45-12-17/FA-45-16-17/FA-45-20-17 Rotors
Products
Product Category
adapters (3)
Manufacturer
Package Size
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
EP5820770007
Adapter for Eppendorf
®
FA-45-12-17/FA-45-16-17/FA-45-20-17 Rotors,
capacity, 1 HPLC tube, pack of 4
Expand
View Pricing
EP5820768002
Adapter for Eppendorf
®
FA-45-12-17/FA-45-16-17/FA-45-20-17 Rotors,
capacity, 1 x 1.5 - 2.0 mL vessels, pack of 4
Expand
View Pricing
EP5820769009
Adapter for Eppendorf
®
FA-45-12-17/FA-45-16-17/FA-45-20-17 Rotors,
capacity, 1 cryotube, pack of 4
Expand
View Pricing