Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Acrylamide/Bis-acrylamide solution
Products
Brand
Sigma-Aldrich (6)
Physical Form
Product Line
Quality Segment
Sterilization Method
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
A3574
Acrylamide/Bis-acrylamide solution,
30%, BioReagent, suitable for electrophoresis, 29:1
Expand
View Pricing
A3699
Acrylamide/Bis-acrylamide solution,
30%, BioReagent, suitable for electrophoresis, 37.5:1
Expand
View Pricing
A9926
Acrylamide/Bis-acrylamide solution,
40%, BioReagent, suitable for electrophoresis, 19:1
Expand
View Pricing
A7802
Acrylamide/Bis-acrylamide solution,
40%, BioReagent, suitable for electrophoresis, 29:1
Expand
View Pricing
A7168
Acrylamide/Bis-acrylamide solution,
40%, BioReagent, suitable for electrophoresis, 37.5:1
Expand
View Pricing
A3449
Acrylamide/Bis-acrylamide solution,
30%, BioReagent, suitable for electrophoresis, 19:1
Expand
View Pricing