Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aconitine
Synonym(s):
Acetylbenzoylaconine
Empirical Formula (Hill Notation):
C
34
H
47
NO
11
CAS Number:
302-27-2
Molecular Weight:
645.74
EC Number:
206-121-7
Products
Product Category
bioactive small molecules (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (2)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A8001
Aconitine,
≥95% (HPLC), crystalline
Expand
View Pricing
PHL89376
Aconitine,
phyproof
®
Reference Substance
Expand
View Pricing
44787
Aconitine,
analytical standard
Expand
View Pricing