Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ace Trubore
®
PTFE stirrer assembly
Products
Manufacturer
Length (in)
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z400521
Ace Trubore
®
PTFE stirrer assembly,
for flask size, 250 ‑ 500 mL, joint: ST/NS 24/40
Expand
View Pricing
Z400548
Ace Trubore
®
PTFE stirrer assembly,
for flask size, 500 ‑ 1,000 mL, joint: ST/NS 29/42
Expand
View Pricing
Z400556
Ace Trubore
®
PTFE stirrer assembly,
for flask size, 2,000 ‑ 3,000 mL, joint: ST/NS 29/42
Expand
View Pricing
Z400564
Ace Trubore
®
PTFE stirrer assembly,
joint: ST/NS 45/50, for flask size, 5,000 ‑ 12,000 mL
Expand
View Pricing