Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ace three-neck round-bottom flasks
Products
Product Category
flasks (8)
Brand
Sigma-Aldrich (5)
Manufacturer
Capacity (mL)
Closure Type
Package Size
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z125776
Ace three-neck round-bottom flasks,
angled, capacity 3,000 mL, Joints: ST/NS 24/40 (3)
Expand
View Pricing
Z125679
Ace three-neck round-bottom flasks,
vertical, capacity 1,000 mL, center joint: ST/NS 29/42, side Joints: ST/NS 24/40 (2)
Expand
View Pricing
Z125695
Ace three-neck round-bottom flasks,
vertical, capacity 5,000 mL, Joints: ST/NS 24/40 (3)
Expand
View Pricing
Z125725
Ace three-neck round-bottom flasks,
angled, capacity 300 mL, Joints: ST/NS 24/40 (3)
Expand
View Pricing
Z125687
Ace three-neck round-bottom flasks,
vertical, capacity 3,000 mL, Joints: ST/NS 24/40 (3)
Expand
View Pricing
Z125784
Ace three-neck round-bottom flasks,
angled, capacity 5,000 mL, Joints: ST/NS 24/40 (3)
Expand
View Pricing
Z125652
Ace three-neck round-bottom flasks,
vertical, capacity 500 mL, center joint: ST/NS 29/42, side Joints: ST/NS 24/40 (2)
Expand
View Pricing
Z125741
Ace three-neck round-bottom flasks,
angled, capacity 500 mL, center joint: ST/NS 29/42, side Joints: ST/NS 24/40 (2)
Expand
View Pricing