Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ace receiving flask adapter
Synonym(s):
Ace receiving flask adapter
Products
Product Category
adapters (4)
Manufacturer
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z682209
Ace receiving flask adapter,
joint: ST/NS 29/32, fits Büchi
®
models R-200/R-205
Expand
View Pricing
Z682187
Ace receiving flask adapter,
joint: ST/NS 24/40, fits Büchi
®
models R-200/R-205
Expand
View Pricing
Z682195
Ace receiving flask adapter,
joint: ST/NS 24/40, fits Büchi
®
models R-114/R-144
Expand
View Pricing
Z682217
Ace receiving flask adapter,
joint: ST/NS 29/42, fits Büchi
®
models R-200/R-205
Expand
View Pricing