Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
81-Well microtube storage box, for 1.5-2.0 mL tubes
Products
Product Category
boxes (8)
Color
Manufacturer
Package Size
Special Grade
Natural (1)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
HS120040
81-Well microtube storage box, for 1.5-2.0 mL tubes,
assorted colors, Blue, Green, Purple, Yellow, Orange, polypropylene
Expand
View Pricing
HS120034
81-Well microtube storage box, for 1.5-2.0 mL tubes,
natural, polypropylene
Expand
View Pricing
HS120041
81-Well microtube storage box, for 1.5-2.0 mL tubes,
black, polypropylene
Expand
View Pricing
HS120038
81-Well microtube storage box, for 1.5-2.0 mL tubes,
yellow, polypropylene
Expand
View Pricing
HS120039
81-Well microtube storage box, for 1.5-2.0 mL tubes,
orange, polypropylene
Expand
View Pricing
HS120035
81-Well microtube storage box, for 1.5-2.0 mL tubes,
blue, polypropylene
Expand
View Pricing
HS120037
81-Well microtube storage box, for 1.5-2.0 mL tubes,
pink, polypropylene
Expand
View Pricing
HS120036
81-Well microtube storage box, for 1.5-2.0 mL tubes,
green, polypropylene
Expand
View Pricing