
6-Beta-Hydroxybudesonide

Synonym(s):
((2R,6aR,6bS,7S,8aS,8bS,11aR,12aS,12bS)-2,7-dihydroxy-8b-(2-hydroxyacetyl)-6a,8a-dimethyl-10-propyl-1,2,6a,6b,7,8,8a,8b,11a,12,12a,12b-dodecahydro-4H-naphtho[2′,1′:4,5]indeno[1,2-d][1,3]dioxol-4-one)
Empirical Formula (Hill Notation):
C25H34O7
CAS Number:
Molecular Weight:
446.53
Products
Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing